Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pli | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001104420.3 | NP_001097890.1 | 6 - 7 | 1123 | 82 | |
NM_079742.4 | NP_524466.1 | 6 - 7 | 1195 | 82 | ||
GenBank mRNA | AF091624.1 | - | 6 - 7 | 1100 | 82 | |
AY069632.1 | - | 6 - 7 | 1171 | 82 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |