Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hr96 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_079769.3 | NP_524493.1 | 1 - 2 | 978 | 79 | |
GenBank mRNA | AY051486.1 | - | 1 - 2 | 960 | 79 | |
U36792.1 | - | 1 - 2 | 936 | 79 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |