Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Stt3B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_079770.3 | NP_524494.1 | 4 - 5 | 2338 | 75 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Drosophila melanogaster |