Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ets97D | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001276071.1 | NP_001263000.1 | 3 - 4 | 371 | 75 | |
NM_079799.3 | NP_524523.2 | 3 - 4 | 371 | 75 | ||
GenBank mRNA | BT025224.2 | - | 3 - 4 | 371 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Drosophila melanogaster |