Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ser | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001300629.1 | NP_001287558.1 | 3 - 4 | 1062 | 72 | |
NM_079803.3 | NP_524527.3 | 3 - 4 | 790 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |