Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Klp98A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001260410.2 | NP_001247339.1 | 5 - 6 | 1970 | 85 | |
NM_079808.4 | NP_524532.2 | 5 - 6 | 1970 | 85 | ||
GenBank mRNA | BT132947.1 | - | 5 - 6 | 1939 | 85 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |