Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ets98B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001300641.1 | NP_001287570.1 | 3 - 4 | 1065 | 69 | |
NM_079811.4 | NP_524535.2 | 3 - 4 | 1541 | 69 | ||
GenBank mRNA | BT053684.1 | - | 3 - 4 | 1513 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Drosophila melanogaster |