Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Or98a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_079812.2 | NP_524536.2 | 1 - 2 | 765 | 117 |
Set Membership: |
Amplicon length greater than or equal to 101 G-protein coupled receptor Drosophila melanogaster |