Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dhc98D | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001170288.1 | NP_001163759.1 | 27 - 28 | 8688 | 105 | |
NM_001276107.1 | NP_001263036.1 | 21 - 22 | 6370 | 105 | ||
NM_079817.3 | NP_524541.3 | 28 - 29 | 8709 | 105 |
Set Membership: |
Amplicon length greater than or equal to 101 Drosophila melanogaster |