Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dhc98D | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001170288.1 | NP_001163759.1 | 44 - 45 | 15458 | 71 | |
NM_001276107.1 | NP_001263036.1 | 38 - 39 | 13140 | 71 | ||
NM_079817.3 | NP_524541.3 | 45 - 46 | 15479 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |