Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cyp4c3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_079859.4 | NP_524598.1 | 6 - 7 | 1121 | 72 | |
GenBank mRNA | BT010108.2 | - | 6 - 7 | 1198 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |