Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CycG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001276217.2 | NP_001263146.1 | 2 - 3 | 911 | 58 | |
NM_079870.4 | NP_524609.2 | 2 - 3 | 911 | 58 | ||
NM_170555.3 | NP_733434.1 | 2 - 3 | 1754 | 58 | ||
NM_170556.3 | NP_733435.1 | 2 - 3 | 1309 | 58 | ||
NM_170557.3 | NP_733436.1 | 2 - 3 | 1167 | 58 | ||
GenBank mRNA | AY058720.1 | - | 2 - 3 | 1760 | 58 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |