Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: faf | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001260458.1 | NP_001247387.1 | 9 - 10 | 6275 | 107 | |
NM_001260459.1 | NP_001247388.1 | 9 - 10 | 6323 | 107 | ||
NM_001260460.2 | NP_001247389.2 | 9 - 10 | 6291 | 107 | ||
NM_001260461.1 | NP_001247390.1 | 10 - 11 | 6389 | 107 | ||
NM_001300698.1 | NP_001287627.1 | 9 - 10 | 6291 | 107 | ||
NM_079873.5 | NP_524612.2 | 9 - 10 | 6291 | 107 | ||
NM_170576.3 | NP_733455.1 | 9 - 10 | 6291 | 107 |
Set Membership: |
Amplicon length greater than or equal to 101 Drosophila melanogaster |