Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sulf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001275697.1 | NP_001262626.1 | 5 - 6 | 2147 | 64 | |
NM_001275698.1 | NP_001262627.1 | 4 - 5 | 1506 | 64 | ||
NM_080248.3 | NP_524987.1 | 4 - 5 | 1940 | 64 | ||
GenBank mRNA | BT120003.1 | - | 4 - 5 | 1935 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |