Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cic | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001104373.2 | NP_001097843.1 | 12 - 13 | 5679 | 73 | |
NM_001260274.2 | NP_001247203.1 | 18 - 19 | 6907 | 73 | ||
NM_001275826.1 | NP_001262755.1 | 17 - 18 | 7644 | 73 | ||
NM_080253.4 | NP_524992.1 | 12 - 13 | 4392 | 73 | ||
GenBank mRNA | AJ252268.1 | - | 12 - 13 | 4370 | 73 | |
BT044183.1 | - | 12 - 13 | 4371 | 73 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Drosophila melanogaster |