Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cic | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001104373.2 | NP_001097843.1 | 3 - 4 | 2349 | 75 | |
NM_001260273.2 | NP_001247202.1 | 3 - 4 | 2349 | 75 | ||
NM_001260274.2 | NP_001247203.1 | 9 - 10 | 4864 | 75 | ||
NM_001260275.2 | NP_001247204.2 | 3 - 4 | 2137 | 75 | ||
NM_001275826.1 | NP_001262755.1 | 8 - 9 | 5586 | 75 | ||
NM_080253.4 | NP_524992.1 | 3 - 4 | 2349 | 75 | ||
GenBank mRNA | AJ252268.1 | - | 3 - 4 | 2327 | 75 | |
BT044183.1 | - | 3 - 4 | 2328 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |