Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dhap-at | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001276027.1 | NP_001262956.1 | 2 - 3 | 845 | 63 | |
NM_001276028.1 | NP_001262957.1 | 2 - 3 | 920 | 63 | ||
NM_080271.4 | NP_525010.1 | 2 - 3 | 920 | 63 | ||
GenBank mRNA | BT099931.1 | - | 2 - 3 | 920 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Drosophila melanogaster |