Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pcmt | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001275358.1 | NP_001262287.1 | 1 - 2 | 194 | 58 | |
NM_080508.4 | NP_536756.1 | 1 - 2 | 194 | 58 | ||
GenBank mRNA | AY070630.1 | - | 1 - 2 | 189 | 58 | |
AY075527.1 | - | 1 - 2 | 189 | 58 | ||
U37432.2 | - | 1 - 2 | 61 | 58 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Drosophila melanogaster |