Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trc8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001276141.1 | NP_001263070.1 | 2 - 3 | 1637 | 65 | |
NM_170413.3 | NP_733292.1 | 2 - 3 | 1389 | 65 | ||
NM_170414.2 | NP_733293.1 | 2 - 3 | 1389 | 65 | ||
NM_206580.2 | NP_996303.1 | 2 - 3 | 1389 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |