Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Atg16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001144652.2 | NP_001138124.2 | 6 - 7 | 1473 | 68 | |
NM_170432.3 | NP_733311.2 | 4 - 5 | 758 | 68 | ||
NM_170433.3 | NP_733312.2 | 3 - 4 | 767 | 68 | ||
NM_170434.4 | NP_733313.2 | 6 - 7 | 1449 | 68 | ||
GenBank mRNA | AY058742.1 | - | 4 - 5 | 758 | 68 | |
BT024267.1 | - | 6 - 7 | 1399 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Drosophila melanogaster |