Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: stops | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001170309.2 | NP_001163780.1 | 8 - 9 | 1448 | 65 | |
NM_170537.2 | NP_733416.2 | 7 - 8 | 1264 | 65 | ||
NM_170538.3 | NP_733417.1 | 8 - 9 | 1403 | 65 | ||
GenBank mRNA | AY047558.1 | - | 7 - 8 | 1020 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Drosophila melanogaster |