Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: dpr11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001275391.1 | NP_001262320.1 | 3 - 4 | 1489 | 70 | |
NM_176409.2 | NP_788586.1 | 3 - 4 | 1489 | 70 | ||
GenBank mRNA | AY121627.1 | - | 3 - 4 | 1489 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |