Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mical | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001260086.3 | NP_001247015.1 | 17 - 18 | 13562 | 65 | |
NM_001260087.3 | NP_001247016.2 | 17 - 18 | 13802 | 65 | ||
NM_176443.2 | NP_788620.1 | 13 - 14 | 7562 | 65 | ||
NM_176444.2 | NP_788621.1 | 15 - 16 | 8366 | 65 | ||
NM_176445.2 | NP_788622.1 | 14 - 15 | 13407 | 65 | ||
NM_176446.2 | NP_788623.1 | 13 - 14 | 13363 | 65 | ||
NM_176447.2 | NP_788624.1 | 17 - 18 | 13529 | 65 | ||
NM_176448.2 | NP_788625.1 | 14 - 15 | 13303 | 65 | ||
NM_176449.3 | NP_788626.2 | 13 - 14 | 13247 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |