Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mical | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001144560.3 | NP_001138032.1 | 2 - 3 | 1811 | 89 | |
NM_001260085.2 | NP_001247014.1 | 4 - 5 | 1858 | 89 | ||
NM_001260086.3 | NP_001247015.1 | 4 - 5 | 1906 | 89 | ||
NM_001260087.3 | NP_001247016.2 | 5 - 6 | 1933 | 89 | ||
NM_001316573.1 | NP_001303502.1 | 5 - 6 | 1933 | 89 | ||
NM_001316574.1 | NP_001303503.1 | 5 - 6 | 1933 | 89 | ||
NM_176443.2 | NP_788620.1 | 5 - 6 | 1933 | 89 | ||
NM_176444.2 | NP_788621.1 | 5 - 6 | 1933 | 89 | ||
NM_176445.2 | NP_788622.1 | 2 - 3 | 1811 | 89 | ||
NM_176446.2 | NP_788623.1 | 1 - 2 | 1767 | 89 | ||
NM_176447.2 | NP_788624.1 | 5 - 6 | 1933 | 89 | ||
NM_176448.2 | NP_788625.1 | 2 - 3 | 1707 | 89 | ||
NM_176449.3 | NP_788626.2 | 2 - 3 | 1720 | 89 | ||
NM_176450.1 | NP_788627.1 | 5 - 6 | 1859 | 89 | ||
GenBank mRNA | AY118290.1 | - | 5 - 6 | 1859 | 89 |
Set Membership: |
Amplicon length between 86 and 100 Drosophila melanogaster |