Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mical | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001260085.2 | NP_001247014.1 | 10 - 11 | 3426 | 74 | |
NM_001260086.3 | NP_001247015.1 | 10 - 11 | 3474 | 74 | ||
NM_001260087.3 | NP_001247016.2 | 10 - 11 | 3432 | 74 | ||
NM_001316573.1 | NP_001303502.1 | 10 - 11 | 3432 | 74 | ||
NM_001316574.1 | NP_001303503.1 | 10 - 11 | 3432 | 74 | ||
NM_176444.2 | NP_788621.1 | 10 - 11 | 3432 | 74 | ||
NM_176445.2 | NP_788622.1 | 7 - 8 | 3310 | 74 | ||
NM_176446.2 | NP_788623.1 | 6 - 7 | 3266 | 74 | ||
NM_176447.2 | NP_788624.1 | 10 - 11 | 3432 | 74 | ||
NM_176448.2 | NP_788625.1 | 7 - 8 | 3206 | 74 | ||
NM_176449.3 | NP_788626.2 | 7 - 8 | 3219 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |