Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cad86C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001260102.1 | NP_001247031.1 | 11 - 12 | 3464 | 108 | |
NM_001260103.1 | NP_001247032.1 | 10 - 11 | 3380 | 108 | ||
NM_001260104.1 | NP_001247033.1 | 11 - 12 | 3482 | 108 | ||
NM_001260105.1 | NP_001247034.1 | 10 - 11 | 4029 | 108 | ||
NM_176458.4 | NP_788635.3 | 11 - 12 | 4120 | 108 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Drosophila melanogaster |