Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GstD3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_176479.2 | NP_788656.1 | 1 | 341 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Drosophila melanogaster |