Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nsf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001300389.1 | NP_001287318.1 | 3 - 4 | 1480 | 87 | |
NM_001300390.1 | NP_001287319.1 | 3 - 4 | 1480 | 87 | ||
NM_176499.2 | NP_788676.1 | 3 - 4 | 1480 | 87 | ||
GenBank mRNA | BT023784.1 | - | 3 - 4 | 1415 | 87 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Drosophila melanogaster |