Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pxb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001275680.1 | NP_001262609.1 | 2 - 3 | 822 | 71 | |
NM_176504.2 | NP_788681.1 | 2 - 3 | 822 | 71 | ||
GenBank mRNA | AB081475.1 | - | 2 - 3 | 704 | 71 | |
BT126212.1 | - | 2 - 3 | 799 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |