Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ppn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001170290.2 | NP_001163761.1 | 9 - 10 | 6966 | 53 | |
NM_176575.3 | NP_788752.2 | 10 - 11 | 7137 | 53 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |