Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AnxB9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001275867.1 | NP_001262796.1 | 4 - 5 | 1020 | 75 | |
NM_001275868.1 | NP_001262797.1 | 4 - 5 | 1021 | 75 | ||
NM_206531.2 | NP_996253.1 | 4 - 5 | 996 | 75 | ||
GenBank mRNA | BT044420.1 | - | 4 - 5 | 995 | 75 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Drosophila melanogaster |