Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Myo95E | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001032035.2 | NP_001027206.1 | 11 - 12 | 3241 | 68 | |
NM_001032037.2 | NP_001027208.1 | 12 - 13 | 3255 | 68 | ||
NM_206555.2 | NP_996278.1 | 11 - 12 | 3225 | 68 | ||
GenBank mRNA | AF454350.1 | - | 11 - 12 | 3225 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |