Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Myo95E | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001032033.2 | NP_001027204.2 | 15 - 16 | 3807 | 147 | |
NM_001032035.2 | NP_001027206.1 | 15 - 16 | 3986 | 147 | ||
NM_001032036.2 | NP_001027207.1 | 15 - 16 | 3823 | 147 | ||
NM_001032037.2 | NP_001027208.1 | 16 - 17 | 4000 | 147 | ||
NM_206555.2 | NP_996278.1 | 15 - 16 | 3970 | 147 | ||
NM_206556.2 | NP_996279.1 | 14 - 15 | 3910 | 147 | ||
GenBank mRNA | AF454350.1 | - | 15 - 16 | 3970 | 147 | |
AF454351.1 | - | 14 - 15 | 3910 | 147 | ||
BT011452.1 | - | 15 - 16 | 3818 | 147 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Drosophila melanogaster |