Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCAP-R | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001382065.1 | NP_001368957.1 | 3 - 4 | 1147 | 82 | |
NM_206574.4 | NP_996297.3 | 3 - 4 | 1147 | 82 | ||
GenBank mRNA | BT133202.1 | - | 3 - 4 | 1147 | 82 |
Set Membership: |
Amplicon length between 71 and 85 G-protein coupled receptor Drosophila melanogaster |