Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SMC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_142954.4 | NP_651211.2 | 3 - 4 | 825 | 90 |
Set Membership: |
Amplicon length between 86 and 100 Drosophila melanogaster |