Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SMC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_142954.4 | NP_651211.2 | 9 - 10 | 3588 | 54 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Drosophila melanogaster |