Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: vis | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_078990.3 | NP_523714.2 | 2 - 3 | 209 | 121 | |
NM_176157.2 | NP_788337.1 | 1 - 2 | 292 | 121 | ||
GenBank mRNA | AY058750.1 | - | 1 - 2 | 483 | 121 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Drosophila melanogaster |