Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CstF50 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_143626.3 | NP_651883.1 | 1 - 2 | 1011 | 109 | |
GenBank mRNA | AY094802.1 | - | 1 - 2 | 939 | 109 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Drosophila melanogaster |