Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Patsas | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001370040.1 | NP_001356957.1 | 1 - 2 | 1789 | 110 | |
NM_001370041.1 | NP_001356894.1 | 1 - 2 | 1789 | 110 | ||
NM_164993.2 | NP_723724.1 | 1 - 2 | 1789 | 110 | ||
GenBank mRNA | AY058389.1 | - | 1 - 2 | 1759 | 110 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Drosophila melanogaster |