Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: vsg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001274717.1 | NP_001261646.1 | 2 - 3 | 195 | 120 | |
NM_168363.2 | NP_729535.1 | 1 - 2 | 301 | 120 | ||
NM_168364.2 | NP_729536.1 | 2 - 3 | 195 | 120 | ||
GenBank mRNA | BT001711.1 | - | 2 - 3 | 183 | 120 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Drosophila melanogaster |