Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Jupiter | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_169404.4 | NP_731595.1 | 2 - 3 | 336 | 112 | |
NM_169405.4 | NP_731596.1 | 2 - 3 | 336 | 112 | ||
NM_169407.4 | NP_731598.1 | 3 - 4 | 400 | 112 | ||
GenBank mRNA | BT006004.1 | - | 3 - 4 | 397 | 112 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Drosophila melanogaster |