Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CrebB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001258811.3 | NP_001245740.1 | 3 - 4 | 1207 | 70 | |
NM_001347813.2 | NP_001334685.1 | 3 - 4 | 783 | 70 | ||
NM_206781.4 | NP_996504.1 | 3 - 4 | 930 | 70 | ||
GenBank mRNA | BT028813.2 | - | 2 - 3 | 498 | 70 | |
BT088933.1 | - | 3 - 4 | 1188 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Drosophila melanogaster |