Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AnxB10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001169333.2 | NP_001162804.1 | 2 | 425 | 90 | |
NM_057267.4 | NP_476615.1 | 2 | 425 | 90 | ||
GenBank mRNA | AY061328.1 | - | 2 | 398 | 90 | |
M34069.1 | - | 2 | 381 | 90 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Recommended Assay Drosophila melanogaster |