Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Acp65Aa | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_057934.3 | NP_477282.2 | 2 | 434 | 75 | |
GenBank mRNA | AY113609.1 | - | 2 | 435 | 75 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Drosophila melanogaster |