Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: a6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001272212.1 | NP_001259141.1 | 2 | 1839 | 97 | |
NM_057961.5 | NP_477309.1 | 2 | 1831 | 97 | ||
GenBank mRNA | AY069426.1 | - | 2 | 1783 | 97 | |
Y16066.1 | - | 1782 | 97 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Recommended Assay Drosophila melanogaster |