Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TotM | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001258958.1 | NP_001245887.1 | 2 | 264 | 69 | |
NM_078758.3 | NP_523482.1 | 2 | 264 | 69 | ||
GenBank mRNA | AY035992.1 | - | 2 | 264 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Drosophila melanogaster |