Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: trn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001274867.1 | NP_001261796.1 | 2 | 3112 | 120 | |
NM_079331.4 | NP_524055.2 | 2 | 3112 | 120 | ||
GenBank mRNA | AY051439.1 | - | 2 | 2921 | 120 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Drosophila melanogaster |