Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GstD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001038953.2 | NP_001034042.1 | 2 | 774 | 117 | |
NM_079602.5 | NP_524326.1 | 2 | 756 | 117 | ||
GenBank mRNA | S51044.1 | - | 1 | 810 | 117 | |
X14233.1 | - | 688 | 117 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Drosophila melanogaster |