Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: inaF-B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001169255.1 | NP_001162726.1 | 2 | 2833 | 143 | |
GenBank mRNA | BT015289.1 | - | 2 | 2810 | 143 |
Gene Symbol: inaF-D | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001103488.2 | NP_001096958.1 | 2 | 2928 | 143 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Drosophila melanogaster |