Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: wus | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001298426.1 | NP_001285355.1 | 2 | 1611 | 141 | |
NM_132952.3 | NP_573180.1 | 2 | 1611 | 141 | ||
GenBank mRNA | AY058516.1 | - | 2 | 1539 | 141 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Drosophila melanogaster |